Sterilising pan, Nalgene®
Supplier: Thermo Fisher Scientific
6910-0618
NALG6910-0618EA 0 SGD
NALG6910-0618
Sterilising pan, Nalgene®
Trays/Dishes/Pots Sterilising Trays/Pans
PP with cover.
  • Cover minimises spillage of hazardous materials
  • Resistant to aggressive disinfectants
  • Can also be used to sterilise labware instruments and utensils
  • Stackable
Allows pipettes to be fully immersed in disinfectant.
Meets OSHA Standard 29 CFR (Part 1910.1030) for use as protection against blood-borne pathogens.
Certifications: Meets OSHA Standard 29 CFR (Part 1910.1030) for use as protection against blood-borne pathogens.