Biohazard transport carriers, Nalgene®
Supplier: Thermo Fisher Scientific
7135-0001
NALG7135-0001EA 158 SGD
NALG7135-0001
Biohazard transport carriers, Nalgene®
Carriers Test Tubes Transport Carriers
Transparent PC with silicone gasket and PC clamps.
  • For 13 or 16 mm Vacutainer test tubes
  • Adaptable for other test tubes
  • Tough and break resistant with excellent visibility of contents
  • Easy to grasp side handles are moulded in
  • Clamps hold the carrier securely closed and ensure leakproof seal
This closed system carrier is designed to protect the lab worker during transportation of tubes filled with potentially hazardous samples. It is autoclavable without removing lid or opening clamps to avoid hazardous exposure. Warning labels are included.