Stoppers, Rubber, with one hole, Nalgene®
Supplier: Thermo Fisher Scientific
DS0396-0080
NALGDS0396-0080EA 0 SGD
NALGDS0396-0080
Stoppers, Rubber, with one hole, Nalgene®
Stoppers Rubber Stoppers
This rubber filter stopper is for use with analytical test filter funnels and filter funnel with clamp. Size: No. 8.