Bubble point test apparatus, Nalgene®
Supplier: Thermo Fisher Scientific
DS0405-0050
NALGDS0405-0050EA 0 SGD
NALGDS0405-0050
Bubble point test apparatus, Nalgene®
Filters Bottle Top Filters
This Bubble point test apparatus is used to ensure product integrity of MF75 Series filter unit or bottle top filters. Tests filter unit membranes without removing the membrane from the filter unit.
  • Cast aluminium
  • Accepts 6 mm int.Ø tubing
Delivery information: Adapters and O-rings included.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement