Magnetic carboy stirrer, Nalgene®
Supplier: Thermo Fisher Scientific
DS2227-0020
NALGDS2227-0020EA 0 SGD
NALGDS2227-0020
Magnetic carboy stirrer, Nalgene®
Stirrers Magnetic Stirrers
For low speed mixing with Nalgene® 10 and 20 litres carboys with 83 mm closures. Note magnetic stirrer shown is not included, needs to be used in conjuction with magnetic stirrer unit to provide stirring action.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Overall dimensions Length: 75 mm
Shaft Length: 588 mm
Shaft Ø: 13 mm
Environmental certification NAR Certificate of Retirement