Conical-bottom centrifuge tube adapters, Nalgene®
Supplier: Thermo Fisher Scientific
DS3147-0050
NALGDS3147-0050EA 0 SGD
NALGDS3147-0050
Conical-bottom centrifuge tube adapters, Nalgene®
Tubes Centrifuge Tubes
PC, autoclavable.
  • These adapters must be used with Nalgene® conical-bottom centrifuge tubes (35 ml)
  • Fits most commonly used low- and high-speed rotors
  • Easily removed from rotors after use