Safety bottle carrier, Nalgene®
Supplier: Thermo Fisher Scientific
DS5996-0871
NALGDS5996-0871EA 0 SGD
NALGDS5996-0871
Safety bottle carrier, Nalgene®
Carriers Bottle Carriers/Containers
PC.
  • Round or square bottles
  • Heat resistant up to 135 °C and autoclavable
  • Handle telescopes into carrier for storage
These bottle carriers are suitable for the transport or storage of up to 8×500 ml bottles.