Self contained biological indicators (SCBI)
Supplier: CROSSTEX
SCS-06
NAMSSCS-06EA 486 SGD
NAMSSCS-06
Self contained biological indicators (SCBI)
Sterilisation Indicators
Self-contained biological indicators (SCBIs) for monitoring steam sterilisation consist of a durable PC vial with cap, a crushable glass ampoule with Trypticase® soy broth (TSB) modified with pH indicator, and a disc inoculated with Geobacillus stearothermophilus spores.
  • Reduced incubation time of 24 hours
  • Visually distinct positive results indicated by the media transitioning from purple to yellow
  • Chemical indicator on label provides instant indication of exposure
Storage condition: 15 to 30 °C
Relative humidity: 20 – 70%
Certifications: More Product Information