Cell analysis slides, Arthur™
Supplier: NANOENTEK
AC0050
NANOAC0050EA 0 SGD
NANOAC0050
Cell analysis slides, Arthur™
Slides Chamber Slides
Arthur™ Cell analysis slides are designed for use with the Arthur™.
  • Assay for cells including GFP&RFP expression, apoptosis, cell viability, cell cycle, and cell counting
  • Eliminates washing steps and cross contamination between samples (fully disposable)
Size: 110×24×1,9 mm
Order Now
SPECIFICATIONS
Environmentally Preferable