Automated cell counter, ADAM™ MC2
Supplier: NANOENTEK
ADAM-MCII
NANOADAM-MCIIEA 0 SGD
NANOADAM-MCII
Automated cell counter, ADAM™ MC2
Cell Counters and Flow Cytometers Automated Cell Counters
ADAM™ MC2 is an automated fluorescence cell counter that performs cell counting and viability measurements at highest accuracy capable of replacing complex flow cytometry workflows. Combined with the disposable micro-fluidic AccuTip, operation is extremely simple and cost-effective. Utilizing the PI (Propidium Iodide) nucleus staining method, most advanced LED optics and CMOS detection technology, ADAM-MC2 is the perfect solution when working with aggregated, irregular shaped or challenging cells like CAR-T cells, CAR-NK cells or adipose-derived stem cells.
  • Reports number of total
More Product Information