Counting chambers, C-Chip
Supplier: NANOENTEK
DHC-B01 DHC-B02 DHC-F01 DHC-M01 DHC-N01 DHC-N04
NANODHC-B01EA 270 SGD
NANODHC-B01 NANODHC-B02 NANODHC-F01 NANODHC-M01 NANODHC-N01 NANODHC-N04
Counting chambers, C-Chip
Slides Counting Chambers
Quartz grade optical plastic counting chambers applicable in blood analysis (hematology).
  • Guaranteed repeatability and reliability, eliminate the need for washing and reuse
  • Disposable hemocytometers that do not need coverslips
  • Chip to reduce exposure to infectious samples and contamination
Suitable for cell counting and culture, concentration measurement and viability test, in bacteria and fungal spore counting, IVF, IUI tasks of sperm counting.
Order Now
SPECIFICATIONS
Environmentally Preferable