Sterilised write-on bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
B01064WA B01195WA B01196WA B01445WA
NASCB01064WAEA 0 SGD
NASCB01064WA NASCB01195WA NASCB01196WA NASCB01445WA
Sterilised write-on bags, Whirl-Pak®
Bags Sample Bags
Blended, virgin polyethylene film with Write-On strip provides exceptional clarity, superior strength, and sterilised after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Convenient write-on strip for easy labeling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured
More Product Information