Hydrated PolyProbe Sampling Bag with Sampling Sponge
Supplier: NASCO FORT ATKINSON
B01592
NASCB01592EA 0 SGD
NASCB01592
Hydrated PolyProbe Sampling Bag with Sampling Sponge
Bags Sample Bags
Make environmental surface sampling faster and easier Write-on label makes identifying samples easy and convenient Each bag contains a polyurethane sponge, premoistened with HiCap neutralizing broth Terminally gamma processed to guarantee sterility -The PolyProbe is sealed in a 24 oz. (710 ml) write-on Whirl-Pak bag and hydrated with 10 ml of HiCap Neutralizing Broth.
ags are Collection broths play a critical role in the success of an environmental monitoring program. HiCap neutralizing broth is the first collection broth developed specifically for use in the food and beverage industry asMore Product Information