Table Top High Speed Micro Centrifuge
Supplier: NEUATION TECHNOLOGIES
IFUGEM15K-B
NEUAIFUGEM15K-BEA 0 SGD
NEUAIFUGEM15K-B
Table Top High Speed Micro Centrifuge
Centrifuges Microcentrifuges
Delivering top-notch accuracy in every cycle despite high load, iFuge M15K has been designed keeping in mind the requirements of the clients. The lightning fast 15000 RPM microprocessor controlled centrifuge can perform at peak efficiency and give the desired results. It gives smooth and silent operation even at the maximum speed.
  • Microprocessor controlled efficient performance
  • Variable speed from 500 to 15000 RPM
  • Intuitive interface for quick and convenient setting with last run memory feature
  • Smooth and silent operation even at maximum speed
  • Large
More Product Information