Ultra-Low temperature freezers, CryoCube F101h
Supplier: NEW BRUNSWICK
F101340001
NEWBF101340001EA 0 SGD
NEWBF101340001
Ultra-Low temperature freezers, CryoCube F101h
Freezers Ultra Low Temperature Freezers
The CryoCube F101h ULT freezers offers a personal storage solution in a convenient package. The compact design means the unit will fit below a standard laboratory bench so samples are always securely close-at-hand. F101h has 2 inner compartments for storing for up to 6 racks/60 boxes giving space for up to 6000 vessels (1,5 ml). These freezers with efficient and reliable 2-stage compressor systems help reduce energy consumption, which helps to lower costs. Energy consumption: (–70/–80 °C): 4,6/6,3 KWh/day.
  • Green cooling liquids (R170/R290) with low Global Warming Potential (GWP)
  • Easy-
More Product Information