Freezers, CryoCube® F740
Supplier: NEW BRUNSWICK
F740320021
NEWBF740320021EA 0 SGD
NEWBF740320021
Freezers, CryoCube® F740
Freezers Ultra Low Temperature Freezers
CryoCube F740 is designed to store more of your samples securely while maintaining optimal energy usage.
  • Polyurethane foam and vacuum insulation panels
  • Led interface
  • Green cooling liquids
  • Air-cooling
  • Easy-to-read, eye level, flush mounted control panel and alarm status
These freezers are equipped with a touchscreen interface where data are stored on-board and can be checked 24/7. If necessary, data as well as service reports can be extracted by a front-mounted USB port. The electronic lock system enables controlled access and higher sample safety.