PIG® BLUE absorbent socks
Supplier: NEW PIG
4048
NEWP4048EA 0 SGD
NEWP4048
PIG® BLUE absorbent socks
Spill Control Materials
Surround spills to prevent spreading. Extra-fine vermiculite filler pulls in liquid and hugs floors to stay in place.
  • Heaviest sock, features extra-dense construction to hug floors and stay in place whilst containing spills
  • Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water
Ideal for diking and damming liquids. Can be air-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable