Spill-X® Portable Chemical Neutralising Kit
Supplier: NEW PIG
GEN522
NEWPGEN522EA 0 SGD
NEWPGEN522
Spill-X® Portable Chemical Neutralising Kit
Spill Control Materials
Neutralises smaller spills for safer cleanup and handling - great for labs. Neutralising capacity up to 1/2 gal. per container or 3 gal. total, 1 each.
  • Solidified liquid is easier to clean up
  • Granular base speeds spill cleanup to limit worker exposure
  • Fluid absorbed: Neutralises acids; neutralises bases; neutralises solvents
Packed with spill control agents that neutralise a wide range of hazardous chemicals and solvents. Portable Kit can be carried right to the spill or wall mounted for extra convenience.
Spill-X-A acid-neutralising adsorbentMore Product Information