PIG® spill response bag
Supplier: NEW PIG
KITE320 KITE220
NEWPKITE320EA 783 SGD
NEWPKITE320 NEWPKITE220
PIG® spill response bag
Spill Control Materials
PIG absorbents in the ideal spill kit bag - hi-viz, water-resistant, and easy to close, hang and carry.
  • Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
  • PIG aborbent socks stop spreading spills; PIG mat absorbs quickly
  • Temporary disposal bags help make cleanup easier
  • Bright-yellow bag with bold Spill Kit lettering for quick identification
  • Strap for easy carrying and hanging
Compact and easy to store kit is great for use in trucks or equipment.
Caution: To ensure effectiveness and your safety, we recommend thatMore Product Information