PIG® universal mat rolls
Supplier: NEW PIG
MAT202-01
NEWPMAT202-01EA 462 SGD
NEWPMAT202-01
PIG® universal mat rolls
Spill Control Materials
Eight layers of 100% PP are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated.
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime so mat stays on the job longer
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
  • Can be wrung out and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable