PIG® universal mats
Supplier: NEW PIG
MAT203 MAT240 MAT204 MAT231 MAT155 MAT251
NEWPMAT203EA 321 SGD
NEWPMAT203 NEWPMAT240 NEWPMAT204 NEWPMAT231 NEWPMAT155 NEWPMAT251
PIG® universal mats
Spill Control Materials
These mats last 2 times longer than ordinary mats so it requires fewer change-outs. Save money with the mat that absorbs anything and works anywhere.
  • Eight layers of 100% PP are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime so mat stays on the job
More Product Information