PIG® 4-in-1® Mat
Supplier: NEW PIG
MAT284
NEWPMAT284EA 0 SGD
NEWPMAT284
PIG® 4-in-1® Mat
Spill Control Materials
This versatile mat roll can also do work as a pad, sock or wipes. You can stock just one absorbent and still handle a variety of leaks, drips and spills.
  • Exclusive 4-in-1 mat is a pad, roll, sock and wipes all in one
  • Use as a pad to catch drips and soak up spills; roll out to cover large areas; fold along score lines to make a thick, absorbent "sock"; tear off a piece to use as a low-lint wipes
  • No matter how you use it, this mat builds on the proven eight-layer, 100% PP construction of PIG Mat
  • Scuff-resistant, spunbond outer layer resists wear and tear,
More Product Information