PIG® mini drip deck
Supplier: NEW PIG
PAK371
NEWPPAK371EA 130 SGD
NEWPPAK371
PIG® mini drip deck
Trays/Dishes/Pots Spillage Trays/Pans
If containers leak, drip or crack, make sure a Mini Drip Deck with built-in sump is in position to catch the liquid.
  • Translucent sidewalls let you easily inspect fluid level in sump
  • Compact size fits on tabletop or counter; ideal for use in labs
  • Pour spout for easy draining
Chemical-resistant polyethylene tray features built-in sump containment to trap leaks and drips whilst you work.
Caution: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of thisMore Product Information