Spill containment trays, SW series
Supplier: NEW PIG
PAKE274SW
NEWPPAKE274SWEA 0 SGD
NEWPPAKE274SW
Spill containment trays, SW series
Trays/Dishes/Pots Spillage Trays/Pans
Lightweight and durable PE spill trays are ideal for storing several small to medium sized containers of liquids.
  • Chemical and rust resistant
  • Stackable for easy transportation and storage
  • Standard trays with high and durable sidewalls; containers sit deep into the sump, minimising any potential splashes
  • Spill trays with grate feature an anti-slip, removable grate that keeps containers away from collected liquids
Caution: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammableMore Product Information