Boiling baths, Clifton
Supplier: NICKEL ELECTRO
1101E 1105E 1026G 1026E 1104E 1101ER 1104ER 1105ER 1026ER
NICK1101EEA 0 SGD
NICK1101E NICK1105E NICK1026G NICK1026E NICK1104E NICK1101ER NICK1104ER NICK1105ER NICK1026ER
Boiling baths, Clifton
Baths Boiling Baths
Fixed temperature baths with concentric ringed lids. The steam produced provides an effective source of heat for samples that may otherwise be thermally damaged by a naked flame. Models 1020G and 1026G are gas baths, which do not have their own electrical power source, they are designed to be heated by a hotplate or gas burner.
  • Choice of round or rectangular baths
  • All electrically heated baths have a 1000 W heater and integral constant level device to regulate the level of water
  • Rings can be used to vary the diameter of the hole in the lid
  • Optional energy
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable