Spoon spatulas, broad shape
Supplier: NICKEL ELECTRO
2002
NICK2002EA 44.3 SGD
NICK2002
Spoon spatulas, broad shape
Spatulas
Pure nickel (99,6%).
  • One end is flat and spatula-shaped, the other end is spoon-shaped
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean