Double spatulas, Chattaway type, stainless steel
Supplier: NICKEL ELECTRO
2315
NICK2315EA 0 SGD
NICK2315
Double spatulas, Chattaway type, stainless steel
Spatulas
Multipurpose spatula made of stainless steel for a wide range of uses. One end is flat and spatula-shaped, the other end is rounded and turned up at 45°. Available in light or medium weight design.
  • Very good chemical resistance
  • Very good thermal resistance
  • Quick and easy to clean