Vortex mixers, Cyclone
Supplier: NICKEL ELECTRO
CM-2
NICKCM-2EA 0 SGD
NICKCM-2
Vortex mixers, Cyclone
Shakers Vortex Shakers
The Cyclone mixers create a vortex in the tube, which gives a highly effective mixing action. The Cyclone+ is designed for tube, micro plate, flask and micro tube mixing at slow speeds using accessory platforms which can be quickly interchanged. It is ideal for pre-treating paraffin specimens for FISH assys, hybridization step, pre-warm the probe to room temperature, vortex and spin down.
  • Variable speed
  • Skirted rubber mixing cup prevents liquid entering the casing
  • Accepts vessels up to 25 mm diameter
  • Continuous running at the chosen speed or on demand by
More Product Information