Panoramic autoclavable goggles, BCAP series, BioClean Clearview™
Supplier: Nitritex
BCAP
NITRBCAPEA 0 SGD
NITRBCAP
Panoramic autoclavable goggles, BCAP series, BioClean Clearview™
Goggles
BioClean Clearview™ autoclavable cleanroom goggles have a super-soft thermoplastic rubber frame to provide wearer comfort.
  • Toughened anti-scratch, anti-fog lens
  • Super-soft frame
  • Indirect ventilation system
  • Ideal for wearing over eyeglasses
  • Non-linting, latex-free headband
  • Tested to withstand 40 autoclave cycles of 30 minutes duration at 121 °C/250 °F (under laboratory conditions)
  • Anti-fog performance remaining for up to 25 cycles with no degradation (under laboratory conditions)
BioClean Clearview™ Autoclavable Panoramic Goggles have a toughened, anti-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable