Marker pens, sterile, BioClean Permaflow™
Supplier: Nitritex
S-BPFP-BK1
NITRS-BPFP-BK1EA 0 SGD
NITRS-BPFP-BK1
Marker pens, sterile, BioClean Permaflow™
Pens Permanent Markers
BioClean Permaflow™ Pens are processed to ensure ISO Class 4 and EU GMP Grade A compatibility and autoseal ink prevents drying.
  • Fade and water resistant
  • Quick drying
  • Non-toxic
Packaging: Ten pieces double bagged