Respirator filter cartridges
Supplier: Honeywell Safety
75SC
NORH75SCEA 0 SGD
NORH75SC
Respirator filter cartridges
Respirators Reusable Respirators Respirator Filters
For use with North safety products air purifying dual cartridge respirators. Adapter assembly attaches pancake filter to a gas and vapour cartridges and attaches to chemical cartridges except for the Defender® multipurpose cartridge.
  • NIOSH approved when used with Nort Safety Products 7700, 7600, 5500, and 5400 air purifying respirators