Neutralising liquids, Spilfyter® Kolorsafe
Supplier: NPS
410001 410004 430004
NPSD410001EA 51.3 SGD
NPSD410001 NPSD410004 NPSD430004
Neutralising liquids, Spilfyter® Kolorsafe
Spill Control Materials
Neutralise hazardous caustic and acid spills (including hydrofluoric acid) with these easy-to-use liquid neutralisers.
  • Liquid formula generates less gas and heat during neutralisation than dry equivalent products
  • For treating spills on vertical or sloping surfaces
  • Kolorsafe color change process which indicates when enough neutraliser has been applied to the spill and spill is safe for absorption and disposal