IVF ICSI dish, Nunc™
Supplier: Thermo Fisher Scientific
150265
NUNC150265EA 0 SGD
NUNC150265
IVF ICSI dish, Nunc™
Culture Dishes Cell Culture Dishes
Non treated surface, sterile (SAL 10⁻⁶), with lid. This IVF ICSI dish meet the exacting requirements of IVF testing, certification, and validation, providing consistently reliable, high quality results during fertilisation.
  • Maintain optimal temperature with true flat bottom
  • Low profile provides optimal angles during ICSI process
  • Clearly see sample with super polished optical centre
  • Increased safety with easy grip design
  • Certificates of conformity available for every lot
Produced under ISO 13485 with full lot traceability
Certifications: ProducedMore Product Information