Chambered cover glasses, Nunc™, Lab-Tek™
Supplier: Thermo Fisher Scientific
171080
NUNC171080EA 0 SGD
NUNC171080
Chambered cover glasses, Nunc™, Lab-Tek™
Cover Glasses
These cover glasses provide secure content on a slide while viewing under a microscope.
For use with slide flask (NUNC170920) and Thermo Scientific™ Flaskette™ products.