Cell culture plates and Multidishes, Nunclon™ Sphera™
Supplier: Thermo Fisher Scientific
174929 174925 174927 174930 174931 174932
NUNC174929EA 0 SGD
NUNC174929 NUNC174925 NUNC174927 NUNC174930 NUNC174931 NUNC174932
Cell culture plates and Multidishes, Nunclon™ Sphera™
Microplates Cell Culture Plates
The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids.
  • Optimises spheroid formation
  • Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
  • Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimises variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth, providing a better modelMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable