Sealing tape for MicroWell™ plates, Nunc™
Supplier: Thermo Fisher Scientific
236366
NUNC236366EA 199 SGD
NUNC236366
Sealing tape for MicroWell™ plates, Nunc™
Seals Microplate Seals
Polyester film with acrylic adhesive. Provides an effective seal for all MicroWell™ plates used in PCR and cell culture.
  • Resistant to temperatures between −70 and +100 °C
  • Minimum evaporation and well-to-well cross-contamination
Dimensions: 134×80 mm
Dimensions: 134×80 mm  
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement