Microplates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
249943 249944 249945 249946 249947 249949 249950 264573 264575 264576 264579 267245 267334 267342 267350 267385
NUNC249943EA 0 SGD
NUNC249943 NUNC249944 NUNC249945 NUNC249946 NUNC249947 NUNC249949 NUNC249950 NUNC264573 NUNC264575 NUNC264576 NUNC264579 NUNC267245 NUNC267334 NUNC267342 NUNC267350 NUNC267385
Microplates, 96- and 384-well, Nunc™
Microplates HTS Microplates
PP, untreated, without lid. These plates are ideal for liquid phase assays, sample preparation, compound storage and chemical synthesis as they are non toxic and resistant to most chemicals, solvents and alcohols. White plates for luminescence applications maximise reflection of signal and minimise autoluminescence. Black plates for fluorescence applications reduce background fluorescence and minimise back-scattered light. Coloured plates are ideal for storage applications.
  • Compatible with robotics and automated systems
  • Choice of conical (V) or round (U)
More Product Information