Plates, Nunc™ OmniTrays
Supplier: Thermo Fisher Scientific
264728
NUNC264728EA 0 SGD
NUNC264728
Plates, Nunc™ OmniTrays
Microplates HTS Microplates
PS, sterile, with lid. Useful for screening libraries and for DNA hybridisation methods, these 90 ml trays are also suitable for standard Petri dish techniques and bacterial growth.
  • Compatible with robotic systems
  • Sturdy wall design facilitates handling and stacking
  • Can be frozen at −20 °C
External dimensions: 128×86 mm
Recommended working volume: 35 ml
Recommended working volume: 35 ml
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement