Racks for cryogenic vials, Nunc™
Supplier: Thermo Fisher Scientific
331825 331826 331827 331828
NUNC331825EA 0 SGD
NUNC331825 NUNC331826 NUNC331827 NUNC331828
Racks for cryogenic vials, Nunc™
Racks Cryogenic Vial Racks
  • Latch rack with standard ANSI microplate footprint
  • Holds 48 internally-threaded CryoTubes™ or Cryobank vials
  • Use pivoting lid on bench or lock it for transport and storage
  • Format facilitates automated picking/placing, capping/decapping, and multi-channel liquid handling
  • Designed for –80°C and liquid nitrogen (vapour phase) storage