Nunc™ Universal Latch Racks for 48 Storage Tubes
Supplier: Thermo Fisher Scientific
331830 331835 331830-BR 331835-BR
NUNC331830EA 0 SGD
NUNC331830 NUNC331835 NUNC331830-BR NUNC331835-BR
Nunc™ Universal Latch Racks for 48 Storage Tubes
Racks Cryogenic Vial Racks
Use versatile Thermo Scientific™ Nunc™ 1,8 ml Externally-threaded universal tubes inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
  • Barcoded rack
  • Sterile
  • Code type: 2D, linear and human readable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10×10 arrays
  • External threads limit contamination
  • Automation-pickable in
More Product Information