CryoFlex™ sleeves for CryoLine™ tubes, Nunc™
Supplier: Thermo Fisher Scientific
343958
NUNC343958EA 0 SGD
NUNC343958
CryoFlex™ sleeves for CryoLine™ tubes, Nunc™
Cryogenic Straws
PE sleeves for storage in liquid nitrogen and to provide extra safety when freezing hazardous samples.
  • Can be cut to the desired length
  • Shrinks with gentle heating
External Ø×L: 14×500 mm