Centrifuge tubes, Nunc™
Supplier: Thermo Fisher Scientific
376813
NUNC376813EA 0 SGD
NUNC376813
Centrifuge tubes, Nunc™
Tubes Centrifuge Tubes
PP, conical bottom, graduated, sterile, autoclavable without cap.
  • Withstands forces up to 7000×g with adapter for 200 ml
  • Moulded graduations