Immunoassay microplates
Supplier: Thermo Fisher Scientific
95029390 3355 3455 3555 3655 3855 6301 6302 6309 6310 6404 6405 9502227 9502887 95029100 95029140 95029180 95029330
NUNC95029390EA 0 SGD
NUNC95029390 NUNC3355 NUNC3455 NUNC3555 NUNC3655 NUNC3855 NUNC6301 NUNC6302 NUNC6309 NUNC6310 NUNC6404 NUNC6405 NUNC9502227 NUNC9502887 NUNC95029100 NUNC95029140 NUNC95029180 NUNC95029330
Immunoassay microplates
Microplates Immunology Microplates and Strips
Designed specifically for immunoassay applications, this range of products have binding surfaces that include Immulon®, universal binding (UB) and enhanced binding (EB). These make good experimental design easier, allowing development of assays with stringent pH, buffer, pKa and resin requirements.
  • Available in 96-well or strip plate format for matching to the requirements of automated instruments
  • Microplates with breakable strips offer maximum efficiency for processing both large and small sample numbers, as the exact number of wells required can be used
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement ISO 14001 (Certificate Registration No. EMS 623400)