UV microplates, Nunc™
Supplier: Thermo Fisher Scientific
8404
NUNC8404EA 0 SGD
NUNC8404
UV microplates, Nunc™
Microplates HTS Microplates
Perform DNA and protein quantitation in 260 nm and 280 nm range with Thermo Scientific™ Nunc™ 96-Well UV microplates. The 96-Well format allows for a higher throughput of sample screening compared to cuvettes.
  • Non sterile
  • Flat bottom (F-well)