DeepWell™ plates, 96 square well, Nunc™
Supplier: Thermo Fisher Scientific
95040452
NUNC95040452EA 0 SGD
NUNC95040452
DeepWell™ plates, 96 square well, Nunc™
Microplates Deep Well Microplates
Ideal for sample collection, storage and combinatorial chemistry library applications.
  • Square well shape reduces liquid retention
  • Resistant to most chemicals, solvents and alcohols used in combinatorial chemistry
  • Sterile format certified free from RNase