Precision balances, Explorer®
Supplier: OHAUS
83021348 83021341 83021350 83021356 83021357 83021360 83021361 83021362 83021365 83021342 30061979 30061980 30061981 30061982 30061983 30061984
OHAU83021348EA 0 SGD
OHAU83021348 OHAU83021341 OHAU83021350 OHAU83021356 OHAU83021357 OHAU83021360 OHAU83021361 OHAU83021362 OHAU83021365 OHAU83021342 OHAU30061979 OHAU30061980 OHAU30061981 OHAU30061982 OHAU30061983 OHAU30061984
Precision balances, Explorer®
Balances and Scales
A range of balances that incorporate a touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mounted on a base, above a base on an optional column, on a wall, or remotely (up to 3 metres distant with an optional cable). The touch screen display, with universal icons and informative prompts, simplifies set-up functions and basic operation. Then the operator has the added choice to use proximity sensors for hands-free operation when handling sensitive samples. These balances have multiple application modes from basic weighing to SQC applications,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable