Analytical balances, Adventurer®, Ohaus
Supplier: OHAUS
30100600 30100602 30100604 30100601 30100603
OHAU30100600EA 0 SGD
OHAU30100600 OHAU30100602 OHAU30100604 OHAU30100601 OHAU30100603
Analytical balances, Adventurer®, Ohaus
Balances and Scales
Balances with multiple features but uncomplicated weighing capabilities provide the user with rapid, accurate results for routine weighing and measurement activities. Applications include weighing, formulation, check weighing and percentage weighing. Ideal for use in laboratories, pharamacies, light industrial areas and in classrooms.
  • Fast stabilisation and reliable operation
  • Wide viewing angle colour touch screen (109 mm) for quick access to it's application modes
  • Dual USB ports with GLP/GMP tracking capabilities
  • Two piece, draught shield side doors
More Product Information