Semi-micro balances, Explorer®
Supplier: OHAUS
30139510 30139511 30139512 30139513 30139514
OHAU30139510EA 0 SGD
OHAU30139510 OHAU30139511 OHAU30139512 OHAU30139513 OHAU30139514
Semi-micro balances, Explorer®
Balances and Scales
These semi-microbalances have been designed and built for sophisticated professional weighing applications where the user requires extreme accuracy.
  • 'High Speed Single Module' weighing cell delivers ultra-fast stabilisation times typically ≤3 seconds for 0,1 mg and ≤8 seconds for 0,01 mg
  • Intuitive software with an icon-based colour touch screen display
  • AutoCal™ automatic internal calibration which uses two internal weights to perform a linear calibration
  • Save data to USB
  • Four touchless sensors allow the operator to perform common functions and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable