Mechanical balances, Harvard Trip®, 1400 and 1500 series
Supplier: OHAUS
80000005
OHAU80000005EA 0 SGD
OHAU80000005
Mechanical balances, Harvard Trip®, 1400 and 1500 series
Balances and Scales
These two pan balances are designed to provide the accuracy necessary in comparative weighing.
Metal base and beam and pan made of stainless steel. Features built-in sliding masses, magnetic damping, self-aligning beam design and floating agate bearings, counterbalancing knob for quick zeroing, hardened steel edges.
Dimensions H×L×W: 160×240×140 mm
Ordering information: Weight set 2000 g model and accessory sold separately.