Bacterial DNA kit, E.Z.N.A.®

Supplier: OMEGA BIO-TEK

D3350-02 D3350-00 D3350-01
OMEGD3350-02EA 0 SGD
OMEGD3350-02 OMEGD3350-00 OMEGD3350-01
Bacterial DNA kit, E.Z.N.A.®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The E.Z.N.A.® bacterial DNA kit allows the rapid and reliable isolation of high quality total cellular DNA from a wide variety of bacterial species. This kit uses optimised lysis condition and up to 1×10⁹ bacterial cells can be processed for each column. There are no organic extractions, thus reducing plastic waste and hands on time to allow multiple samples to be processed in parallel. Bacterial cells are grown to log-phase and harvested. The cell wall is removed by lysozyme digestion and bead beating, followed by protease digestion. Following lysis, binding conditions are adjusted and the sample is applied to a HiBind DNA spin-column. Two rapid wash steps remove trace salts and protein contaminants, and DNA is finally eluted in water or elution buffer. Purified DNA can be directly used in downstream applications without the need for further purification.


  • Suitable for most bacterial strains
  • DNA isolation in less than 20 minutes (after lysis)
  • Optimised buffers guarantee pure DNA
  • No organic extractions
  • Purified DNA suitable for most applications

Order Now

SPECIFICATIONS

Description E.Z.N.A.® Bacterial DNA Mini Kit
Environmentally Preferable
Application
Target molecule DNA
Refine target molecule Bacterial DNA
Sample type Bacteria
Method / Format Column (silica-based)
Prep size Mini
Sample size 3 ml log phase culture

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR